Ethyl-3-chloropropionate ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
C1274-25G C1274-100G
TCIAC1274-25GEA 112 EUR
TCIAC1274-25G TCIAC1274-100G
Ethyl-3-chloropropionate ≥97.0% (by GC)
Ethyl-3-chloropropionate
Formule: C₅H₉ClO₂
Kookpunt: 162°C
Vlampunt: 54°C
MDL Nummer: MFCD00000989
CAS nummer: 623-71-2
EINECS: 210-809-2
UN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=519c6a70d21de5d9e2a95c245d6db7ee2793a9e4&vid=519c6a70d21de5d9e2a95c245d6db7ee2793a9e4&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %