1-Boc-4-hydroxypiperidine ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: N-Boc-4-piperidinol, tert-butyl 4-hydroxypiperidine-1-carboxylate, N-BOC-4-Hydroxypiperidine, tert-Butyl 4-hydroxypiperidine-1-carboxylate

B2671-25G B2671-5G
TCIAB2671-25GEA 115 EUR
TCIAB2671-25G TCIAB2671-5G
1-Boc-4-hydroxypiperidine ≥97.0% (by GC)
1-Boc-4-hydroxypiperidine
Formule: C₁₀H₁₉NO₃
MDL Nummer: MFCD01075174
CAS nummer: 109384-19-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=887aabf3d4213f1a72865a06c49f97e484b9ace0&vid=887aabf3d4213f1a72865a06c49f97e484b9ace0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 59.0 to 66.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR