α-Acetyl-γ-butyrolactone ≥99.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
alpha-Acetyl-gamma-hydroxybutyric acid Lactone, α-Acetyl-γ-hydroxybutyric acid Lactone, 3-Acetyldihydrofuran-2(3H)-one, 2-Acetyl-γ-butyrolactone, 2-Acetylbutyrolactone, alpha-Acetyl-gamma-butyrolactone, α-Acetylbutyrolactone, 3-Acetyl-4,5-dihydro-2(3H)-furanone
A0681-25G
A0681-500G
TCIAA0681-25GEA
17.6
EUR
TCIAA0681-25G
TCIAA0681-500G
α-Acetyl-γ-butyrolactone ≥99.0% (by GC)
α-Acetyl-γ-butyrolactone
Formule:
C₆H₈O₃ Vlampunt: 135°C |
MDL Nummer:
MFCD00005394 CAS nummer: 517-23-7 EINECS: 208-235-2 Merck Index: 14,00083 |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...