2,4,6-Trimethylpyrylium tetrafluoroborate ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

T2375-25G T2375-5G
TCIAT2375-25GEA 259 EUR
TCIAT2375-25G TCIAT2375-5G
2,4,6-Trimethylpyrylium tetrafluoroborate ≥98.0% (by HPLC, titration analysis)
2,4,6-Trimethylpyrylium tetrafluoroborate
Formule: C₈H₁₁BF₄O
Smeltpunt: 208°C
MDL Nummer: MFCD00032098
CAS nummer: 773-01-3
EINECS: 212-256-2
UN: 1759
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c785959cdcf3cf8ae86df539c999a80474059ec&vid=9c785959cdcf3cf8ae86df539c999a80474059ec&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR