Cyclopentaan ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Pentamethylene
C0699-25ML
C0699-500ML
C0699-100ML
TCIAC0699-25MLEA
17.6
EUR
TCIAC0699-25ML
TCIAC0699-500ML
TCIAC0699-100ML
Cyclopentaan ≥98.0% (by GC)
Cyclopentaan
Formule:
C₅H₁₀ Kookpunt: 50°C Dichtheid: 0,746 g/cm³ (20 °C) Vlampunt: -41 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00001356 CAS nummer: 287-92-3 EINECS: 206-016-6 UN: 1146 ADR: 3,II Merck Index: 14,02741 |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...