(Bromomethyl)cyclohexane ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
B1708-25G B1708-500G B1708-100G
TCIAB1708-25GEA 62.7 EUR
TCIAB1708-25G TCIAB1708-500G TCIAB1708-100G
(Bromomethyl)cyclohexane ≥97.0% (by GC)
(Bromomethyl)cyclohexane
Formule: C₇H₁₃Br
MW: 177.08 g/mol
Kookpunt: 94°C
Vlampunt: 70°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001509
CAS nummer: 2550-36-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=816f58abbb60b121f3d0cc1d008a9aa6dfe7a6a7&vid=816f58abbb60b121f3d0cc1d008a9aa6dfe7a6a7&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %