2-Heptylcyclopentanone ≥99.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

H0569-25ML H0569-500ML
TCIAH0569-25MLEA 250 EUR
TCIAH0569-25ML TCIAH0569-500ML
2-Heptylcyclopentanone ≥99.0% (by GC)
2-Heptylcyclopentanone
Formule: C₁₂H₂₂O
MW: 182.31 g/mol
Kookpunt: 264°C
MDL Nummer: MFCD00019315
CAS nummer: 137-03-1
EINECS: 205-273-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f86355186d5ad629720162181239103635ddf96d&vid=f86355186d5ad629720162181239103635ddf96d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR