2-Methylcyclohexanon ≥96.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

M0197-25ML M0197-500ML M0197-100ML
TCIAM0197-25MLEA 31.9 EUR
TCIAM0197-25ML TCIAM0197-500ML TCIAM0197-100ML
2-Methylcyclohexanon ≥96.0% (by GC)
2-Methylcyclohexanon
Formule: C₇H₁₂O
Kookpunt: 165°C
Smeltpunt: –14 °C
Dichtheid: 0,924 g/cm³ (20 °C)
Vlampunt: 48°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001635
CAS nummer: 583-60-8
EINECS: 209-513-6
UN: 2297
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef6e2cc50168ef11a67b41a83effac2186af1e04&vid=ef6e2cc50168ef11a67b41a83effac2186af1e04&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR