2-Acetylcyclohexanone ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 2-Acetyl-1-cyclohexanone

A0881-10ML A0881-25ML
TCIAA0881-10MLEA 57.2 EUR
TCIAA0881-10ML TCIAA0881-25ML
2-Acetylcyclohexanone ≥97.0% (by GC)
2-Acetylcyclohexanone
Formule: C₈H₁₂O₂
Kookpunt: 111°C
Dichtheid: 1,063 g/cm³ (20 °C)
Vlampunt: 79°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001633
CAS nummer: 874-23-7
EINECS: 212-858-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=939ce7fc3ddd1ee68853576819f4b0b257d17651&vid=939ce7fc3ddd1ee68853576819f4b0b257d17651&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Brown clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR