1-Methylcyclopentanol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

M0729-10G M0729-25G
TCIAM0729-10GEA 36.3 EUR
TCIAM0729-10G TCIAM0729-25G
1-Methylcyclopentanol ≥98.0% (by GC)
1-Methylcyclopentanol
Formule: C₆H₁₂O
Kookpunt: 135 °C (1013 hPa)
Smeltpunt: 32…33 °C
Dichtheid: 0,905 g/cm³ (23.5 °C)
Vlampunt: 40°C
MDL Nummer: MFCD00001364
CAS nummer: 1462-03-9
EINECS: 215-963-4
UN: 1325
ADR: 4.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13ba18005082773522d5ec1281daceaca2489d06&vid=13ba18005082773522d5ec1281daceaca2489d06&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR