tert-Butylcyclohexane ≥99.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B0823-25ML B0823-100ML
TCIAB0823-25MLEA 42.9 EUR
TCIAB0823-25ML TCIAB0823-100ML
tert-Butylcyclohexane ≥99.0% (by GC)
tert-Butylcyclohexane
Formule: C₁₀H₂₀
Kookpunt: 171°C
Smeltpunt: –41 °C
Dichtheid: 0,8127 g/cm³ (20 °C)
Vlampunt: 42°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001471
CAS nummer: 3178-22-1
EINECS: 221-652-4
UN: 3295
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca171776c33af9adcdd20b2319094726af23ec9d&vid=ca171776c33af9adcdd20b2319094726af23ec9d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR