Cycloheptene ≥96.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
C0688-25ML C0688-5ML
TCIAC0688-25MLEA 241 EUR
TCIAC0688-25ML TCIAC0688-5ML
Cycloheptene ≥96.0% (by GC)
Cycloheptene
Formule: C₇H₁₂
Kookpunt: 115°C
Smeltpunt: –56 °C
Dichtheid: 0,824 g/cm³ (20 °C)
MDL Nummer: MFCD00004156
CAS nummer: 628-92-2
EINECS: 211-060-4
UN: 2242
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07be40630d09be7cd15f8ee7a83420ff0b5db464&vid=07be40630d09be7cd15f8ee7a83420ff0b5db464&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %