Cyclopentene ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
C0511-25ML
C0511-500ML
C0511-100ML
TCIAC0511-25MLEA
24.2
EUR
TCIAC0511-25ML
TCIAC0511-500ML
TCIAC0511-100ML
Cyclopentene ≥98.0% (by GC)
Cyclopentene
Formule:
C₅H₈ MW: 68.12 g/mol Kookpunt: 46°C Smeltpunt: –135°C Vlampunt: –29°C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00001394 CAS nummer: 142-29-0 EINECS: 205-532-9 UN: 2246 ADR: 3 II |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...