Cyclopentene 95,0 GC_ASSAY_METHOD
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
C1617-25ML C1617-500ML C1617-100ML
TCIAC1617-25MLEA 22 EUR
TCIAC1617-25ML TCIAC1617-500ML TCIAC1617-100ML
Cyclopentene 95,0 GC_ASSAY_METHOD
Cyclopentene
Formule: C₅H₈
MW: 68.12 g/mol
Kookpunt: 46°C
Smeltpunt: –135°C
Vlampunt: –29°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00001394
CAS nummer: 142-29-0
EINECS: 205-532-9
UN: 2246
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=363852c3d08ff04265daea3e31cd2d498860ba51&vid=363852c3d08ff04265daea3e31cd2d498860ba51&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %