2-Bromofluorene ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
B3069-100G
B3069-25G
B3069-5G
TCIAB3069-100GEA
237
EUR
TCIAB3069-100G
TCIAB3069-25G
TCIAB3069-5G
2-Bromofluorene ≥98.0% (by GC)
2-Bromofluorene
Formule:
C₁₃H₉Br MW: 245.12 g/mol Kookpunt: 185°C |
MDL Nummer:
MFCD00001115 CAS nummer: 1133-80-8 EINECS: 214-480-6 |
|
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 112.0 to 116.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...