Myristoyl chloride ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
T0086-25G
T0086-400G
TCIAT0086-25GEA
30.8
EUR
TCIAT0086-25G
TCIAT0086-400G
Myristoyl chloride ≥98.0% (by GC, titration analysis)
Myristoyl chloride
Formule:
C₁₄H₂₇ClO Kookpunt: 165°C |
MDL Nummer:
MFCD00000741 CAS nummer: 112-64-1 EINECS: 203-994-6 UN: 3265 ADR: 8,III |
|
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...