5-Aminopicolinic acid ≥98.0% (by HPLC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 5-Aminopyridine-2-carboxylic acid, 5-Amino-2-pyridinecarboxylic acid
A2386-1G A2386-5G
TCIAA2386-1GEA 53.9 EUR
TCIAA2386-1G TCIAA2386-5G
5-Aminopicolinic acid ≥98.0% (by HPLC)
5-Aminopicolinic acid
Formule: C₆H₆N₂O₂
MW: 138.13 g/mol
Smeltpunt: 218°C
MDL Nummer: MFCD02684600
CAS nummer: 24242-20-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=46cf91f03b866344d3ddad82c471cdca25f5bdbb&vid=46cf91f03b866344d3ddad82c471cdca25f5bdbb&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Light yellow to Light orange powder to crystalline
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 95.0 %