Diethyl phenylmalonate ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

P0311-25ML P0311-500ML
TCIAP0311-25MLEA 17.6 EUR
TCIAP0311-25ML TCIAP0311-500ML
Diethyl phenylmalonate ≥97.0% (by GC)
Diethyl phenylmalonate
Formule: C₁₃H₁₆O₄
Kookpunt: 285°C
MDL Nummer: MFCD00009144
CAS nummer: 83-13-6
EINECS: 201-456-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b66d37705f05ee27544899bcc21af402953368b7&vid=b66d37705f05ee27544899bcc21af402953368b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR