Ethyl glycolate ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
G0242-100G
G0242-25G
TCIAG0242-100GEA
217
EUR
TCIAG0242-100G
TCIAG0242-25G
Ethyl glycolate ≥98.0% (by GC)
Ethyl glycolate
Formule:
C₄H₈O₃ Kookpunt: 160°C Vlampunt: 62°C |
MDL Nummer:
MFCD00021970 CAS nummer: 623-50-7 EINECS: 210-798-4 |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...