Diethyl benzylmalonate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

B0423-25ML B0423-500ML
TCIAB0423-25MLEA 49.5 EUR
TCIAB0423-25ML TCIAB0423-500ML
Diethyl benzylmalonate ≥98.0% (by GC)
Diethyl benzylmalonate
Formule: C₁₄H₁₈O₄
Kookpunt: 300°C
MDL Nummer: MFCD00009166
CAS nummer: 607-81-8
EINECS: 210-142-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df3d8a6fd62e7a02e02901018edb559d212ffa11&vid=df3d8a6fd62e7a02e02901018edb559d212ffa11&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR