Girard's reagent T (carbazoylmethyltrimethylammonium chloride) ≥98.0% (by titrimetric analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
2-Hydrazinyl-N,N,N-trimethyl-2-oxoethanaminium chloride
B0457-25G
B0457-500G
TCIAB0457-25GEA
22
EUR
TCIAB0457-25G
TCIAB0457-500G
Girard's reagent T (carbazoylmethyltrimethylammonium chloride) ≥98.0% (by titrimetric analysis)
Girard's reagent T (carbazoylmethyltrimethylammonium chloride)
Formule:
C₅H₁₄ClN₃O MW: 167.64 g/mol |
MDL Nummer:
MFCD00012009 CAS nummer: 123-46-6 EINECS: 204-629-3 Merck Index: 14,04429 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Argentmetric Titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...