Hydrazinium monobromide ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: Hydrazine monohydrobromide

H0173-25G H0173-500G
TCIAH0173-25GEA 23.1 EUR
TCIAH0173-25G TCIAH0173-500G
Hydrazinium monobromide ≥98.0% (by titrimetric analysis)
Hydrazinium monobromide
Formule: H₅BrN₂
MDL Nummer: MFCD00065394
CAS nummer: 13775-80-9
EINECS: 237-412-7
UN: 3290
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48a7c4cbfbb9e50054af3b42dd530131c898f540&vid=48a7c4cbfbb9e50054af3b42dd530131c898f540&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Purity (Potassium iodate Method) min. 98.0 %
Melting point 86.0 to 90.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR