3-Aminocrotononitrile ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 3-Iminobutyronitrile, 3-Amino-2-butenenitrile, Diacetonitrile, 3-Aminobut-2-enenitrile

A0944-25G A0944-100G
TCIAA0944-25GEA 33 EUR
TCIAA0944-25G TCIAA0944-100G
3-Aminocrotononitrile ≥98.0% (by GC)
3-Aminocrotononitrile
Formule: C₄H₆N₂
MW: 82.11 g/mol
MDL Nummer: MFCD00008071
CAS nummer: 1118-61-2
EINECS: 214-266-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9b54ba6a86f7e7f46b712a4e9f030cc4d992404&vid=c9b54ba6a86f7e7f46b712a4e9f030cc4d992404&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystaline
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR