(±)-Ethyl-2-bromobutyrate ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Ethyl 2-bromobutyrate, Ethyl 2-bromobutanoate
B0800-25G
B0800-500G
TCIAB0800-25GEA
17.6
EUR
TCIAB0800-25G
TCIAB0800-500G
(±)-Ethyl-2-bromobutyrate ≥98.0% (by GC)
(±)-Ethyl-2-bromobutyrate
Formule:
C₆H₁₁BrO₂ MW: 195.06 g/mol Vlampunt: 73°C |
MDL Nummer:
MFCD00000155 CAS nummer: 533-68-6 EINECS: 208-574-6 |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...