1-Butoxy-2,3-epoxypropaan ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
B0697-25ML
B0697-500ML
TCIAB0697-25MLEA
18.7
EUR
TCIAB0697-25ML
TCIAB0697-500ML
1-Butoxy-2,3-epoxypropaan ≥98.0% (by GC)
1-Butoxy-2,3-epoxypropaan
Formule:
C₇H₁₄O₂ Vlampunt: 59°C |
MDL Nummer:
MFCD00005145 CAS nummer: 2426-08-6 EINECS: 219-376-4 UN: 3271 ADR: 3,III |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...