Pentaethylene glycol ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 3,6,9,12-Tetraoxatetradecane-1,14-diol

P1154-25G P1154-5G
TCIAP1154-25GEA 258 EUR
TCIAP1154-25G TCIAP1154-5G
Pentaethylene glycol ≥95.0% (by GC)
Pentaethylene glycol
Formule: C₁₀H₂₂O₆
Kookpunt: 184°C
MDL Nummer: MFCD00002878
CAS nummer: 4792-15-8
EINECS: 225-341-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ff82f2d09b7d5478d487afd7d38fc06d1e3fde1&vid=9ff82f2d09b7d5478d487afd7d38fc06d1e3fde1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR