Diethyl acetylenedicarboxylate ≥96.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
A0089-25ML A0089-250ML
TCIAA0089-25MLEA 81.4 EUR
TCIAA0089-25ML TCIAA0089-250ML
Diethyl acetylenedicarboxylate ≥96.0% (by GC)
Diethyl acetylenedicarboxylate
Formule: C₈H₁₀O₄
Kookpunt: 115°C
Smeltpunt: 1°C
Vlampunt: 94°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00009186
CAS nummer: 762-21-0
EINECS: 212-095-8
UN: 1760
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=197bd469369b29b61d0142ed5c6dc8f11e28d44a&vid=197bd469369b29b61d0142ed5c6dc8f11e28d44a&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 96.0 %