2-Arsanilic acid ≥98.0% (by titrimetric analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
A0529-25G
A0529-5G
TCIAA0529-25GEA
139
EUR
TCIAA0529-25G
TCIAA0529-5G
2-Arsanilic acid ≥98.0% (by titrimetric analysis)
2-Arsanilic acid
Formule:
C₆H₈AsNO₃ |
MDL Nummer:
MFCD00007630 CAS nummer: 2045-00-3 EINECS: 218-064-5 UN: 3465 ADR: 6.1,III |
|
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 150.0 to 155.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...