4-Arsanilic acid ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: p-Arsanilic acid

A0530-25G A0530-500G
TCIAA0530-25GEA 19.8 EUR
TCIAA0530-25G TCIAA0530-500G
4-Arsanilic acid ≥98.0% (by HPLC, titration analysis)
4-Arsanilic acid
Formule: C₆H₈AsNO₃
MDL Nummer: MFCD00007819
CAS nummer: 98-50-0
EINECS: 202-674-3
UN: 3465
ADR: 6.1,III
Merck Index: 14,00792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f43ec2c935b4771a11f63daf0edffcbca1805c7&vid=4f43ec2c935b4771a11f63daf0edffcbca1805c7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in hot Methanol almost transparency
Solubility in hot Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR