5,6-Dihydro-2H-pyran-2-one ≥95.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
D2261-1G D2261-25G D2261-5G
TCIAD2261-1GEA 57.2 EUR
TCIAD2261-1G TCIAD2261-25G TCIAD2261-5G
5,6-Dihydro-2H-pyran-2-one ≥95.0% (by GC)
5,6-Dihydro-2H-pyran-2-one
Formule: C₅H₆O₂
Kookpunt: 117°C
Vlampunt: 46°C
MDL Nummer: MFCD00010439
CAS nummer: 3393-45-1
UN: 1224
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12d7580115e5c706d1a17f4126060265125e57cc&vid=12d7580115e5c706d1a17f4126060265125e57cc&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %