Diphenyl chlorophosphate ≥95.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Diphenyl phosphorochloridate
D1059-25G D1059-500G D1059-100G
TCIAD1059-25GEA 19.8 EUR
TCIAD1059-25G TCIAD1059-500G TCIAD1059-100G
Diphenyl chlorophosphate ≥95.0% (by GC)
Diphenyl chlorophosphate
Formule: C₁₂H₁₀ClO₃P
MW: 268.64 g/mol
Kookpunt: 316°C
MDL Nummer: MFCD00003030
CAS nummer: 2524-64-3
EINECS: 219-759-6
UN: 3265
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5abe6e49bf56eac9643a513587319cc394791431&vid=5abe6e49bf56eac9643a513587319cc394791431&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %