Ethyl-5-bromonicotinate ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
E0866-25G E0866-5G
TCIAE0866-25GEA 196 EUR
TCIAE0866-25G TCIAE0866-5G
Ethyl-5-bromonicotinate ≥98.0% (by GC)
Ethyl-5-bromonicotinate
Formule: C₈H₈BrNO₂
Kookpunt: 92°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00040366
CAS nummer: 20986-40-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a76e1bc42a8574e616fe4ce01cb5215c52677132&vid=a76e1bc42a8574e616fe4ce01cb5215c52677132&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Brown to Dark purple powder to crystal
Purity (GC) min. 98.0 %
Melting point 37.0 to 41.0 deg-C