2-Aminoindan ≥99.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

A1320-25G A1320-5G
TCIAA1320-25GEA 214 EUR
TCIAA1320-25G TCIAA1320-5G
2-Aminoindan ≥99.0% (by GC, titration analysis)
2-Aminoindan
Formule: C₉H₁₁N
Kookpunt: 229°C
MDL Nummer: MFCD00082597
CAS nummer: 2975-41-9
EINECS: 221-021-3
UN: 2735
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1f3ab88e25acec7cea5a000a3904eec1de600fe3&vid=1f3ab88e25acec7cea5a000a3904eec1de600fe3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Purple to Brown clear liquid
Purity (GC) min. 99.0 %
Purity (Neutralization titration) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR