1-Chloronaphthalene ≥85.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Triisodecyl phosphite, α-Chloronaphthalene
C0212-25G C0212-500G C0212-100G
TCIAC0212-25GEA 23.1 EUR
TCIAC0212-25G TCIAC0212-500G TCIAC0212-100G
1-Chloronaphthalene ≥85.0% (by GC)
1-Chloronaphthalene
Formule: C₁₀H₇Cl
MW: 162.62 g/mol
Kookpunt: 263°C
Smeltpunt: –20°C
Vlampunt: 121°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003874
CAS nummer: 90-13-1
EINECS: 201-967-3
UN: 3082
ADR: 9 III
Merck Index: 14,02149
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da25412a8f7eb954e2111451072883555ce1689e&vid=da25412a8f7eb954e2111451072883555ce1689e&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 85.0 %