1,4-Dimethylnaphthalene ≥96.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

D1006-5G D1006-25G
TCIAD1006-5GEA 71.5 EUR
TCIAD1006-5G TCIAD1006-25G
1,4-Dimethylnaphthalene ≥96.0% (by GC)
1,4-Dimethylnaphthalene
Formule: C₁₂H₁₂
Kookpunt: 268°C
Dichtheid: 1,016 g/cm³ (20 °C)
Vlampunt: 128°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004037
CAS nummer: 571-58-4
EINECS: 209-335-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ed7ca9e2c980e2e66bb37b6f42f2e85090672565&vid=ed7ca9e2c980e2e66bb37b6f42f2e85090672565&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR