2,7-Dimethylnaphthalene ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D0752-1G
D0752-5G
TCIAD0752-1GEA
119
EUR
TCIAD0752-1G
TCIAD0752-5G
2,7-Dimethylnaphthalene ≥98.0% (by GC)
2,7-Dimethylnaphthalene
Formule:
C₁₂H₁₂ MW: 156.23 g/mol Kookpunt: 263°C |
MDL Nummer:
MFCD00004121 CAS nummer: 582-16-1 EINECS: 209-477-1 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 95.0 to 98.0 deg-C |
Solubility in Toluene | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...