2,6-Dibromopyridine ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
D1555-25G D1555-250G
TCIAD1555-25GEA 33 EUR
TCIAD1555-25G TCIAD1555-250G
2,6-Dibromopyridine ≥98.0% (by GC)
2,6-Dibromopyridine
Formule: C₅H₃Br₂N
Kookpunt: 249°C
Smeltpunt: 116…119 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006223
CAS nummer: 626-05-1
EINECS: 210-926-9
UN: 2811
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=92743c6aa7617c16cb650348690c334e7ec82181&vid=92743c6aa7617c16cb650348690c334e7ec82181&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 117.0 to 120.0 deg-C
Solubility in hot Methanol almost transparency