2,5-Dichloropyridine ≥99.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
D1607-25G
D1607-5G
TCIAD1607-25GEA
94.6
EUR
TCIAD1607-25G
TCIAD1607-5G
2,5-Dichloropyridine ≥99.0% (by GC)
2,5-Dichloropyridine
Formule:
C₅H₃Cl₂N Kookpunt: 98°C Smeltpunt: 65 °C |
MDL Nummer:
MFCD00006239 CAS nummer: 16110-09-1 EINECS: 240-278-2 ADR: 6.1,III |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 60.0 to 63.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...