3-Chloropyridine ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

C0280-25G C0280-500G C0280-100G
TCIAC0280-25GEA 24.2 EUR
TCIAC0280-25G TCIAC0280-500G TCIAC0280-100G
3-Chloropyridine ≥95.0% (by GC)
3-Chloropyridine
Formule: C₅H₄ClN
Kookpunt: 148°C
Dichtheid: 1,194 g/cm³ (20 °C)
Vlampunt: 66°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006375
CAS nummer: 626-60-8
EINECS: 210-955-7
ADR: 3,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ffdb6c54d1330d274d565c822dea59ac6e5c2795&vid=ffdb6c54d1330d274d565c822dea59ac6e5c2795&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR