2,6-Dichloro-3-nitropyridine ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

D2761-25G D2761-5G
TCIAD2761-25GEA 81.4 EUR
TCIAD2761-25G TCIAD2761-5G
2,6-Dichloro-3-nitropyridine ≥98.0% (by GC)
2,6-Dichloro-3-nitropyridine
Formule: C₅H₂Cl₂N₂O₂
MW: 192.99 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006234
CAS nummer: 16013-85-7
EINECS: 240-151-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84e7083f9e8f20d4394f852b9bd16c6b089cb1da&vid=84e7083f9e8f20d4394f852b9bd16c6b089cb1da&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 63.0 to 66.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR