2,3,5,6-Tetrafluoropyridine ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
T1778-1G T1778-5G
TCIAT1778-1GEA 27.5 EUR
TCIAT1778-1G TCIAT1778-5G
2,3,5,6-Tetrafluoropyridine ≥98.0% (by GC)
2,3,5,6-Tetrafluoropyridine
Formule: C₅HF₄N
Kookpunt: 101°C
Dichtheid: 1,499 g/cm³ (20 °C)
Vlampunt: 31°C
MDL Nummer: MFCD00011734
CAS nummer: 2875-18-5
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d55713640ba5670d7432905212b3f20d59d1c49a&vid=d55713640ba5670d7432905212b3f20d59d1c49a&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %