2,6-Difluoropyridine ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
D1741-25G D1741-5G
TCIAD1741-25GEA 92.4 EUR
TCIAD1741-25G TCIAD1741-5G
2,6-Difluoropyridine ≥98.0% (by GC)
2,6-Difluoropyridine
Formule: C₅H₃F₂N
Kookpunt: 125°C
Dichtheid: 1,26 g/cm³ (20 °C)
Vlampunt: 33°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006227
CAS nummer: 1513-65-1
EINECS: 216-152-8
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ddf93ae2a0bd8c3e5f50e53d8e44c83f1aef056&vid=8ddf93ae2a0bd8c3e5f50e53d8e44c83f1aef056&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %