4-Methoxypyridine ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
M1696-25G M1696-5G
TCIAM1696-25GEA 176 EUR
TCIAM1696-25G TCIAM1696-5G
4-Methoxypyridine ≥98.0% (by GC, titration analysis)
4-Methoxypyridine
Formule: C₆H₇NO
Kookpunt: 190°C
Smeltpunt: 4°C
Vlampunt: 74°C
MDL Nummer: MFCD00674049
CAS nummer: 620-08-6
EINECS: 210-624-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8bea1a64bf22e647f7a2b6ece5ea7a0a072f8262&vid=8bea1a64bf22e647f7a2b6ece5ea7a0a072f8262&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %