2-Chloor-3-methylpyridine ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 2-Chloro-3-methylpyridine, 2-Chloro 3-methylpyridine, 2-Chloro-β-picoline

C1885-25G C1885-5G
TCIAC1885-25GEA 113 EUR
TCIAC1885-25G TCIAC1885-5G
2-Chloor-3-methylpyridine ≥98.0% (by GC)
2-Chloor-3-methylpyridine
Formule: C₆H₆ClN
MW: 127.57 g/mol
Kookpunt: 193°C
Vlampunt: 79°C
MDL Nummer: MFCD00234177
CAS nummer: 18368-76-8
EINECS: 242-242-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6516724904491274b4eabc35e9b023327800fe38&vid=6516724904491274b4eabc35e9b023327800fe38&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR