2-Chloor-6-methylpyridine ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
6-Chloro-2-picoline
C1654-25G
C1654-5G
TCIAC1654-25GEA
170
EUR
TCIAC1654-25G
TCIAC1654-5G
2-Chloor-6-methylpyridine ≥98.0% (by GC)
2-Chloor-6-methylpyridine
Formule:
C₆H₆ClN MW: 127.57 g/mol Kookpunt: 88°C Vlampunt: 73°C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00006245 CAS nummer: 18368-63-3 EINECS: 242-241-6 |
|
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...