2-Picoline ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
2-Methylpyridine, α-Picoline
P0415-25ML
P0415-500ML
TCIAP0415-25MLEA
19.8
EUR
TCIAP0415-25ML
TCIAP0415-500ML
2-Picoline ≥98.0% (by GC, titration analysis)
2-Picoline
Formule:
C₆H₇N Kookpunt: 128°C Smeltpunt: –70 °C Dichtheid: 0,944 g/cm³ (20 °C) Vlampunt: 25°C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00006332 CAS nummer: 109-06-8 EINECS: 203-643-7 UN: 2313 ADR: 3,III Merck Index: 14,07400 |
|
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...