3-Picoline ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
β-Picoline, 3-Methylpyridine
P0416-25ML
P0416-500ML
TCIAP0416-25MLEA
39.6
EUR
TCIAP0416-25ML
TCIAP0416-500ML
3-Picoline ≥98.0% (by GC)
3-Picoline
Formule:
C₆H₇N Kookpunt: 144°C Smeltpunt: –18°C Dichtheid: 0,957 g/cm³ (20 °C) Vlampunt: 36°C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00006402 CAS nummer: 108-99-6 EINECS: 203-636-9 UN: 2313 ADR: 3,III Merck Index: 14,07401 |
|
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...