4-Picoline ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

P0417-25ML P0417-500ML
TCIAP0417-25MLEA 17.6 EUR
TCIAP0417-25ML TCIAP0417-500ML
4-Picoline ≥98.0% (by GC, titration analysis)
4-Picoline
Formule: C₆H₇N
Kookpunt: 144°C
Smeltpunt: 4°C
Dichtheid: 0,9547 g/cm³ (20 °C)
Vlampunt: 37°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006440
CAS nummer: 108-89-4
EINECS: 203-626-4
UN: 2313
ADR: 3,III
Merck Index: 14,07402
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=402f294dfaadae030425c8ae49733cefc27a33f4&vid=402f294dfaadae030425c8ae49733cefc27a33f4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR