3,4-Lutidine ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
L0066-25ML
L0066-500ML
TCIAL0066-25MLEA
19.8
EUR
TCIAL0066-25ML
TCIAL0066-500ML
3,4-Lutidine ≥98.0% (by GC)
3,4-Lutidine
Formule:
C₇H₉N Kookpunt: 179°C Smeltpunt: -6°C Dichtheid: 0,955 g/cm³ (20 °C) Vlampunt: 61°C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00006403 CAS nummer: 583-58-4 EINECS: 209-511-5 UN: 2810 ADR: 6.1,II |
|
Specification Test Results
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...