2,5-Dibromothiophene ≥95.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
D1549-25G D1549-100G
TCIAD1549-25GEA 31.9 EUR
TCIAD1549-25G TCIAD1549-100G
2,5-Dibromothiophene ≥95.0% (by GC)
2,5-Dibromothiophene
Formule: C₄H₂Br₂S
Kookpunt: 210°C
Smeltpunt: –6 °C
Dichtheid: 2,1531 g/cm³ (13.8 °C)
Vlampunt: 99°C
MDL Nummer: MFCD00005420
CAS nummer: 3141-27-3
EINECS: 221-547-3
UN: 2810
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cac2d020992a77f84bc9d29dfb69d4af471e1392&vid=cac2d020992a77f84bc9d29dfb69d4af471e1392&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity (GC) min. 95.0 %